Learn More
Cole-Parmer™ Essentials Suspension Clamp, Nickel-Plated Zinc
Provide support for your specific laboratory equipment with this Nickel-Plated Zinc Suspension Clamp.
Brand: Cole-Parmer™ Essentials 08027-08
Additional Details : Weight : 0.80000kg
Description
- Ideal for suspending thermometers, potash bulbs or drying tubes.
- Integrated connector included.
Specifications
1 Ea. | |
Nickel-plated Zinc | |
Ideal for suspending thermometers, potash bulbs or drying tubes. Integrated connector included | |
136.91 mm |
Hose and Tubing Clamp | |
Integrated Connector | |
5.39 in. | |
Fixed |