Learn More
2-Methylbutyraldehyde, 95%, Thermo Scientific Chemicals
Click to view available options
Quantity:
10 g
25 g
100 g
Description
2-Methylbutyraldehyde is used for lift in woody and cocoa topnote complexes. And also used in cocoa, coffee, wine and nut flavors for lift.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
2-Methylbutyraldehyde is used for lift in woody and cocoa topnote complexes. And also used in cocoa, coffee, wine and nut flavors for lift.
Solubility
Soluble in water, ether, and alcohol.
Notes
Air Sensitive. Store away from strong oxidizing agents. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
2-Methylbutyraldehyde is used for lift in woody and cocoa topnote complexes. And also used in cocoa, coffee, wine and nut flavors for lift.
Solubility
Soluble in water, ether, and alcohol.
Notes
Air Sensitive. Store away from strong oxidizing agents. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
Specifications
Boiling Point | 94°C to 96°C |
Flash Point | 4°C (39°F) |
Quantity | 10 g |
UN Number | UN1989 |
Beilstein | 1633540 |
Formula Weight | 86.13 |
Percent Purity | 95% |
Chemical Name or Material | 2-Methylbutyraldehyde |