missing translation for 'onlineSavingsMsg'
Learn More

2-Fluoro-6-(trifluoromethyl)benzaldehyde, 98%, Thermo Scientific™

Catalog Number p-4812802
Click to view available options
Quantity:
1 g
5 g
This item is not returnable. View return policy
60611-24-7
C8H4F4O
192.11
MFCD00061273
FAKUGVHRTLCKHB-UHFFFAOYSA-N
2-fluoro-6-trifluoromethyl benzaldehyde, benzaldehyde, 2-fluoro-6-trifluoromethyl, alpha,alpha,alpha,6-tetrafluoro-o-tolualdehyde, pubchem4236, 2-fluoro-6-trifluoromethyl-benzaldehyde, acmc-1b5by, otf-bad-6f, 3-fluoro-2-formylbenzotrifluoride, attercop-chm at111495
521933
2-fluoro-6-(trifluoromethyl)benzaldehyde
FC1=CC=CC(=C1C=O)C(F)(F)F
This item is not returnable. View return policy
60611-24-7
C8H4F4O
192.11
MFCD00061273
FAKUGVHRTLCKHB-UHFFFAOYSA-N
2-fluoro-6-trifluoromethyl benzaldehyde, benzaldehyde, 2-fluoro-6-trifluoromethyl, alpha,alpha,alpha,6-tetrafluoro-o-tolualdehyde, pubchem4236, 2-fluoro-6-trifluoromethyl-benzaldehyde, acmc-1b5by, otf-bad-6f, 3-fluoro-2-formylbenzotrifluoride, attercop-chm at111495
521933
2-fluoro-6-(trifluoromethyl)benzaldehyde
FC1=CC=CC(=C1C=O)C(F)(F)F
Boiling Point 156°C
Flash Point 82°C
Assay Percent Range 97.5% min. (GC)
Packaging Glass Bottle
Quantity 1 g
Formula Weight 192.11
Percent Purity 98%
Chemical Name or Material 2-Fluoro-6-(trifluoromethyl)benzaldehyde
compliance-icons
Product Identifier
  • 2-Fluoro-6-(trifluoromethyl)benzaldehyde
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'