Learn More
IBA Solutions For Life Science 2'-Fluoro-2'-dCTP
A modified nucleoside-triphosphate (3´-modified oligos) that prevents nuclease degradation of RNAs.
Brand:  IBA Solutions For Life Science 5-0611-286
Additional Details : Weight : 0.00050kg
 View more versions of this product
Product Code. 15623628
  • 1418.00€ /
Estimated Shipment date
from Supplier 17-06-2020
Add to basket
Description
Description
IBA has a nucleoside-triphosphate synthesis facility that offers an extensive range of triphosphates for many applications.
  • All products are double HPLC purified, desalted and well characterized by HPLC and 31P-NMR
  • They are also available as monophosphates and nucleosides
Specifications
Specifications
Yes
Prevents nuclease degradation of RNAs
10.0 μmol
Yes
Nucleoside-triphosphate