missing translation for 'onlineSavingsMsg'
Learn More

2-(Ethoxymethyl)furan, 98%, Thermo Scientific™

Catalog Number 15505966
Click to view available options
Quantity:
1 g
5 g
This item is not returnable. View return policy
6270-56-0
C7H10O2
126.16
BHGBNDNKYPEAAT-UHFFFAOYSA-N
2-ethoxymethyl furan, ethyl furfuryl ether, furan, 2-ethoxymethyl, furfuryl ethyl ether, 2-furfuryl ethyl ether, unii-9y4ozi563u, ethylfurfuryl ether, 2-ethoxymethyl-furan, furan,2-ethoxymethyl
80455
2-(ethoxymethyl)furan
CCOCC1=CC=CO1
This item is not returnable. View return policy
6270-56-0
C7H10O2
126.16
BHGBNDNKYPEAAT-UHFFFAOYSA-N
2-ethoxymethyl furan, ethyl furfuryl ether, furan, 2-ethoxymethyl, furfuryl ethyl ether, 2-furfuryl ethyl ether, unii-9y4ozi563u, ethylfurfuryl ether, 2-ethoxymethyl-furan, furan,2-ethoxymethyl
80455
2-(ethoxymethyl)furan
CCOCC1=CC=CO1
Boiling Point 150.0°C
Flash Point 41°C
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (GC)
Packaging Glass Bottle
Quantity 1 g
Formula Weight 126.16
Percent Purity 98%
Chemical Name or Material 2-(Ethoxymethyl)furan
compliance-icons compliance-icons
Product Identifier
  • 2-(Ethoxymethyl)furan, 98%
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Flammable liquid Category 3
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H226-Flammable liquid and vapour.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'