Learn More
(±)-2-chlorophenylglycine, 98%, ACROS Organics™
Brand: Acros Organics 433341000
Additional Details : CAS : 88744-36-9 Weight : 0.15000kg
Quantity | 100g |
---|
Chemical Identifiers
88744-36-9 | |
185.61 | |
2-amino-2-2-chlorophenyl acetic acid, 2-2-chlorophenyl glycine, 2-chlorophenylglycine, dl-2-2-chlorophenyl glycine, amino 2-chlorophenyl acetic acid, amino-2-chloro-phenyl-acetic acid, dl-2-chlorophenylglycine, 2-chlorophenyl glycine, 2-chloro-dl-phenylglycine, h-dl-phg 2-cl-oh | |
2-amino-2-(2-chlorophenyl)acetic acid |
C8H8ClNO2 | |
LMIZLNPFTRQPSF-UHFFFAOYSA-N | |
290729 | |
C1=CC=C(C(=C1)C(C(=O)O)N)Cl |
Specifications
(±)-2-chlorophenylglycine | |
White | |
100g | |
97.5 | |
97.5% min. (HPLC) | |
2-amino-2-2-chlorophenyl acetic acid, 2-2-chlorophenyl glycine, 2-chlorophenylglycine, dl-2-2-chlorophenyl glycine, amino 2-chlorophenyl acetic acid, amino-2-chloro-phenyl-acetic acid, dl-2-chlorophenylglycine, 2-chlorophenyl glycine, 2-chloro-dl-phenylglycine, h-dl-phg 2-cl-oh | |
C1=CC=C(C(=C1)C(C(=O)O)N)Cl | |
185.61 | |
185.61 | |
98% |
Authentic | |
182.9°C to 183°C | |
88744-36-9 | |
100.0 | |
C8H8ClNO2 | |
LMIZLNPFTRQPSF-UHFFFAOYSA-N | |
2-amino-2-(2-chlorophenyl)acetic acid | |
290729 | |
Powder |
Safety and Handling
- (±)-2-Chlorophenylglycine
Signal Word
- Warning
Hazard Category
- Serious eye damage/eye irritation Category 2
- Skin corrosion/irritation Category 2
- Specific target organ toxicity Category 3
Hazard Statement
- H319-Causes serious eye irritation.
- H335-May cause respiratory irritation.
- H315-Causes skin irritation.
Precautionary Statement
- P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P302+P352-IF ON SKIN: Wash with plenty of water/.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.