missing translation for 'onlineSavingsMsg'
Learn More

2-Chloro-4-fluoro-5-nitrophenol, 98%, Thermo Scientific™

Catalog Number 15505986
Click to view available options
Quantity:
1 g
5 g
25 g
This item is not returnable. View return policy
84478-75-1
C6H3ClFNO3
191.54
MFCD02670258
NAWVMCKMQMJQMF-UHFFFAOYSA-N
2-chloro-4-fluoro-5-nitrobenzenol, phenol, 2-chloro-4-fluoro-5-nitro, 2-chloro-4-fluoro-5-nitro-phenol, pubchem2849, chlorofluoronitrobenzenol, acmc-1bkdg, ksc495s7h, 2-fluoro-4-chloro-5-hydroxynitrobenzene, 4-chloro-2-fluoro-5-hydroxy-1-nitrobenzene
158655
2-chloro-4-fluoro-5-nitrophenol
OC1=CC(=C(F)C=C1Cl)[N+]([O-])=O
This item is not returnable. View return policy
84478-75-1
C6H3ClFNO3
191.54
MFCD02670258
NAWVMCKMQMJQMF-UHFFFAOYSA-N
2-chloro-4-fluoro-5-nitrobenzenol, phenol, 2-chloro-4-fluoro-5-nitro, 2-chloro-4-fluoro-5-nitro-phenol, pubchem2849, chlorofluoronitrobenzenol, acmc-1bkdg, ksc495s7h, 2-fluoro-4-chloro-5-hydroxynitrobenzene, 4-chloro-2-fluoro-5-hydroxy-1-nitrobenzene
158655
2-chloro-4-fluoro-5-nitrophenol
OC1=CC(=C(F)C=C1Cl)[N+]([O-])=O
Melting Point 99.0°C to 101.0°C
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (GC)
Packaging Glass Bottle
Quantity 1 g
Formula Weight 191.55
Percent Purity 98%
Chemical Name or Material 2-Chloro-4-fluoro-5-nitrophenol
compliance-icons
Product Identifier
  • 2-Chloro-4-fluoro-5-nitrophenol, 98%
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H332-Harmful if inhaled.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'