missing translation for 'onlineSavingsMsg'
Learn More

2-Bromo-6-(hydroxymethyl)pyridine, 96%, Thermo Scientific Chemicals

Catalog Number p-3770745
Click to view available options
Quantity:
1 g
5 g
Packaging:
Glass bottle
This item is not returnable. View return policy
33674-96-3
C6H6BrNO
188.02
MFCD04039884
XDDGKNRSCDEWBR-UHFFFAOYSA-N
6-bromopyridin-2-yl methanol, 2-bromo-6-pyridinemethanol, 2-bromo-6-hydroxymethyl pyridine, 6-bromo-2-hydroxymethyl pyridine, 2-pyridinemethanol, 6-bromo, 6-bromopyridine-2-methanol, 6-bromo-pyridine-2-yl methanol, 6-bromo-2-pyridinemethanol, 6-bromo-pyridin-2-yl-methanol, 6-bromo-pyridin-2-yl methanol
3517811
(6-bromopyridin-2-yl)methanol
OCC1=CC=CC(Br)=N1
This item is not returnable. View return policy
33674-96-3
C6H6BrNO
188.02
MFCD04039884
XDDGKNRSCDEWBR-UHFFFAOYSA-N
6-bromopyridin-2-yl methanol, 2-bromo-6-pyridinemethanol, 2-bromo-6-hydroxymethyl pyridine, 6-bromo-2-hydroxymethyl pyridine, 2-pyridinemethanol, 6-bromo, 6-bromopyridine-2-methanol, 6-bromo-pyridine-2-yl methanol, 6-bromo-2-pyridinemethanol, 6-bromo-pyridin-2-yl-methanol, 6-bromo-pyridin-2-yl methanol
3517811
(6-bromopyridin-2-yl)methanol
OCC1=CC=CC(Br)=N1

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
CAS Min % 95.0
CAS Max % 100.0
Melting Point 34°C to 39°C
Color White
Boiling Point 246°C
Flash Point >110°C
Infrared Spectrum Authentic
Assay Percent Range 95% min. (GC)
Packaging Glass bottle
Quantity 1 g
Formula Weight 188.02
Percent Purity 96%
Physical Form Powder
Chemical Name or Material 2-Bromo-6-(hydroxymethyl)pyridine
Show More Show Less
compliance-icons
Product Identifier
  • 2-Bromo-6-(hydroxymethyl)pyridine
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H312-Harmful in contact with skin.
  • H332-Harmful if inhaled.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'