missing translation for 'onlineSavingsMsg'
Learn More

2-Amino-4-bromophenol, 98%, Thermo Scientific™

Catalog Number p-7037140
Click to view available options
Quantity:
250 mg
1 g
5 g
This item is not returnable. View return policy
40925-68-6
C6H6BrNO
188.02
MFCD00235171
JHRIPENGTGSNPJ-UHFFFAOYSA-N
phenol, 2-amino-4-bromo, 5-bromo-2-hydroxyaniline, 4-bromo-2-aminophenol, 2-amino-4-bromo-phenol, 2-amino-4-bromophenole, 2-amino-4bromophenol, pubchem4511, acmc-209jho, 2-amino4-bromophenol, ksc493s8p
351840
2-amino-4-bromophenol
NC1=CC(Br)=CC=C1O
This item is not returnable. View return policy
40925-68-6
C6H6BrNO
188.02
MFCD00235171
JHRIPENGTGSNPJ-UHFFFAOYSA-N
phenol, 2-amino-4-bromo, 5-bromo-2-hydroxyaniline, 4-bromo-2-aminophenol, 2-amino-4-bromo-phenol, 2-amino-4-bromophenole, 2-amino-4bromophenol, pubchem4511, acmc-209jho, 2-amino4-bromophenol, ksc493s8p
351840
2-amino-4-bromophenol
NC1=CC(Br)=CC=C1O

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
Melting Point 130°C to 135°C
Quantity 250 mg
Beilstein 3236448
Formula Weight 188.02
Percent Purity 98%
Chemical Name or Material 2-Amino-4-bromophenol
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'