missing translation for 'onlineSavingsMsg'
Learn More

2-Amino-4,5-dihydro-1,3-thiazol-4-one hydrochloride, Thermo Scientific™

Catalog Number p-2697722
Click to view available options
Quantity:
1 g
10 g
This item is not returnable. View return policy
2192-06-5
C3H4ClN2OS
151.59
CJPDNRMSRPXGQO-UHFFFAOYSA-N
2-aminothiazol-4 5h-one hydrochloride, 2-aminothiazolinone hcl, 2-amino-4,5-dihydro-1,3-thiazol-4-one hydrochloride, 2-aminothiazolinone hydrochloride, 2-amino-1,3-thiazol-4 5h-one hydrochloride, 4 5h-thiazolone, 2-amino-hydrochloride, 2-amino-1,3-thiazolin-4-one, chloride, 2-aminothiazole-4-one hydrochloride, 2-amino-1,3-thiazol-4-one hydrochloride, c3h4n2os.hcl
2782348
2-amino-1,3-thiazol-4-one;hydrochloride
C1C(=O)N=C(S1)N.Cl
This item is not returnable. View return policy
2192-06-5
C3H4ClN2OS
151.59
CJPDNRMSRPXGQO-UHFFFAOYSA-N
2-aminothiazol-4 5h-one hydrochloride, 2-aminothiazolinone hcl, 2-amino-4,5-dihydro-1,3-thiazol-4-one hydrochloride, 2-aminothiazolinone hydrochloride, 2-amino-1,3-thiazol-4 5h-one hydrochloride, 4 5h-thiazolone, 2-amino-hydrochloride, 2-amino-1,3-thiazolin-4-one, chloride, 2-aminothiazole-4-one hydrochloride, 2-amino-1,3-thiazol-4-one hydrochloride, c3h4n2os.hcl
2782348
2-amino-1,3-thiazol-4-one;hydrochloride
C1C(=O)N=C(S1)N.Cl
CAS Min % 97.0
Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 1 g
Formula Weight 152.6
Percent Purity 97%
Chemical Name or Material 2-Amino-4, 5-dihydro-1, 3-thiazol-4-one hydrochloride
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'