Learn More
2-Acetamido-5-bromopyridine, 98%, ACROS Organics™
Brand: Acros Organics 442420050
Additional Details : CAS : 7169-97-3 Weight : 0.05500kg
Quantity | 5g |
---|
Chemical Identifiers
7169-97-3 | |
215.05 | |
2-acetamido-5-bromopyridine, n-5-bromopyridin-2-yl acetamide, 2-acetylamino-5-bromopyridine, acetamide, n-5-bromo-2-pyridinyl, n-5-bromo-2-pyridyl acetamide, n-5-bromo-2-pyridinyl acetamide, n-5-bromo-pyridin-2-yl-acetamide, n-acetylamino-5-bromopyridine, pubchem5546 | |
N-(5-bromopyridin-2-yl)acetamide |
C7H7BrN2O | |
MJFCOXATGBYERZ-UHFFFAOYSA-N | |
293097 | |
CC(=O)NC1=NC=C(C=C1)Br |
Specifications
2-Acetamido-5-bromopyridine | |
5g | |
97.5% min. (HPLC) | |
2-acetamido-5-bromopyridine, n-5-bromopyridin-2-yl acetamide, 2-acetylamino-5-bromopyridine, acetamide, n-5-bromo-2-pyridinyl, n-5-bromo-2-pyridyl acetamide, n-5-bromo-2-pyridinyl acetamide, n-5-bromo-pyridin-2-yl-acetamide, n-acetylamino-5-bromopyridine, pubchem5546 | |
CC(=O)NC1=NC=C(C=C1)Br | |
215.05 | |
215.05 |
176°C to 178°C | |
7169-97-3 | |
C7H7BrN2O | |
MJFCOXATGBYERZ-UHFFFAOYSA-N | |
N-(5-bromopyridin-2-yl)acetamide | |
293097 | |
98% |
Safety and Handling
- 2-Acetamido-5-bromopyridine
Signal Word
- Warning
Hazard Category
- Acute toxicity Category 4
- Serious eye damage/eye irritation Category 2
- Skin corrosion/irritation Category 2
- Specific target organ toxicity Category 3
Hazard Statement
- H315-Causes skin irritation.
- H319-Causes serious eye irritation.
- H335-May cause respiratory irritation.
- H302-Harmful if swallowed.
Precautionary Statement
- P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
- P302+P352-IF ON SKIN: Wash with plenty of water/.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.