missing translation for 'onlineSavingsMsg'
Learn More

2,5-Dimethyl-1,3-oxazole-4-carboxylic acid, 97%, Thermo Scientific™

Catalog Number p-2697185
Click to view available options
Quantity:
250 mg
1 g
5 g
This item is not returnable. View return policy
23000-14-8
C6H7NO3
141.126
MFCD03011595
LHGRUGVXZLHYKE-UHFFFAOYSA-N
2,5-dimethyloxazole-4-carboxylic acid, dimethyl-1,3-oxazole-4-carboxylic acid, 4-oxazolecarboxylic acid, 2,5-dimethyl, 2,5-dimethyl-1,3-oxazole-4-carboxylicacid, 4-carboxy-2,5-dimethyl-1,3-oxazole, 2,5-dimethyl-oxazole-4-carboxylic acid, 2.5-dimethyl-1,3-oxazole-4-carboxylic acid
2795465
2,5-dimethyl-1,3-oxazole-4-carboxylic acid
CC1=C(N=C(O1)C)C(=O)O
This item is not returnable. View return policy
23000-14-8
C6H7NO3
141.126
MFCD03011595
LHGRUGVXZLHYKE-UHFFFAOYSA-N
2,5-dimethyloxazole-4-carboxylic acid, dimethyl-1,3-oxazole-4-carboxylic acid, 4-oxazolecarboxylic acid, 2,5-dimethyl, 2,5-dimethyl-1,3-oxazole-4-carboxylicacid, 4-carboxy-2,5-dimethyl-1,3-oxazole, 2,5-dimethyl-oxazole-4-carboxylic acid, 2.5-dimethyl-1,3-oxazole-4-carboxylic acid
2795465
2,5-dimethyl-1,3-oxazole-4-carboxylic acid
CC1=C(N=C(O1)C)C(=O)O
CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 250 mg
Formula Weight 141.13
Percent Purity 97%
Chemical Name or Material 2, 5-Dimethyl-1, 3-oxazole-4-carboxylic acid
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'