Learn More
2,5-Dichlorobenzyl bromide, 97%, Acros Organics™
Brand: Acros Organics 443690050
Additional Details : CAS : 85482-13-9 Weight : 0.00500kg Transport : UN number : 3261 Chem class : 8 Pack group : II
Quantity | 5g |
---|
Chemical Identifiers
85482-13-9 | |
239.921 | |
2,5-dichlorobenzyl bromide, 2-bromomethyl-1,4-dichlorobenzene, 2,5-dichlorobenzylbromide, 1-bromomethyl-2,5-dichlorobenzene, alpha-bromo-2,5-dichlorotoluene, benzene, 2-bromomethyl-1,4-dichloro, 2-bromomethyl-1,4-dichloro-benzene, pubchem10208, acmc-1bjf0, 2,5-di-chloro-benzyl bromide | |
2-(bromomethyl)-1,4-dichlorobenzene |
C7H5BrCl2 | |
UUVDOPTUDWJHFK-UHFFFAOYSA-N | |
3495744 | |
C1=CC(=C(C=C1Cl)CBr)Cl |
Specifications
2,5-Dichlorobenzyl bromide | |
134.0°C to 138.0°C (14.0 Torr) | |
5g | |
C7H5BrCl2 | |
UUVDOPTUDWJHFK-UHFFFAOYSA-N | |
2-(bromomethyl)-1,4-dichlorobenzene | |
3495744 | |
97% |
Authentic | |
41.0°C to 43.0°C | |
85482-13-9 | |
2,5-dichlorobenzyl bromide, 2-bromomethyl-1,4-dichlorobenzene, 2,5-dichlorobenzylbromide, 1-bromomethyl-2,5-dichlorobenzene, alpha-bromo-2,5-dichlorotoluene, benzene, 2-bromomethyl-1,4-dichloro, 2-bromomethyl-1,4-dichloro-benzene, pubchem10208, acmc-1bjf0, 2,5-di-chloro-benzyl bromide | |
C1=CC(=C(C=C1Cl)CBr)Cl | |
239.921 | |
239.93 |
Safety and Handling
- 2,5-Dichlorobenzyl bromide, 97%
Signal Word
- Danger
Hazard Category
- Serious eye damage/eye irritation Category 1
- Skin corrosion/irritation Category 1 B
Hazard Statement
- H318-Causes serious eye damage.
- H314-Causes severe skin burns and eye damage.
Precautionary Statement
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
- P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P310-Immediately call a POISON CENTER/doctor/.