Learn More
2,5-Dibromothiazole, 97%, Thermo Scientific Chemicals
Catalog Number p-3757579
25.40 EUR valid until 2024-03-29
Use promo code "21615" to get your promotional price.
Click to view available options
Quantity:
1 g
5 g
Packaging:
Glass bottle
4175-78-4
MFCD00016891
XIBIQFJKUZZLLX-UHFFFAOYSA-N
2,5-dibromothiazole, thiazole, 2,5-dibromo, 2,5-dibormothiazole, thiazole,5-dibromo, 2,5-dibromo-thiazole, pubchem20949, acmc-209jlm, ksc237s3r, #
312394
2,5-dibromo-1,3-thiazole
C1=C(SC(=N1)Br)Br
4175-78-4
MFCD00016891
XIBIQFJKUZZLLX-UHFFFAOYSA-N
2,5-dibromothiazole, thiazole, 2,5-dibromo, 2,5-dibormothiazole, thiazole,5-dibromo, 2,5-dibromo-thiazole, pubchem20949, acmc-209jlm, ksc237s3r, #
312394
2,5-dibromo-1,3-thiazole
C1=C(SC(=N1)Br)Br
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
TRUSTED_SUSTAINABILITY
CAS Min % 96.0
CAS Max % 100.0
Melting Point 45°C to 49°C
Color White to Brown
Infrared Spectrum Authentic
Assay Percent Range 96% min. (GC)
Packaging Glass bottle
Molecular Formula C3HBr2NS
Quantity 1 g
Molecular Weight (g/mol) 242.91
Formula Weight 242.91
Percent Purity 97%
Physical Form Crystalline Powder
Chemical Name or Material 2, 5-Dibromothiazole
Show More Show Less
compliance-icons
Product Identifier
  • 2,5-Dibromothiazole
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.