missing translation for 'onlineSavingsMsg'
Learn More

2-(4-Fluorophenoxy)benzyl alcohol, 95%, Thermo Scientific™

Catalog Number p-7031860
Click to view available options
Quantity:
250 mg
1 g
This item is not returnable. View return policy
70764-39-5
C13H11FO2
218.227
MFCD02082002
LHUOIWVHXSPAQX-UHFFFAOYSA-N
2-4-fluorophenoxy phenyl methanol, 2-4-fluorophenoxy benzyl alcohol
3700253
[2-(4-fluorophenoxy)phenyl]methanol
C1=CC=C(C(=C1)CO)OC2=CC=C(C=C2)F
This item is not returnable. View return policy
70764-39-5
C13H11FO2
218.227
MFCD02082002
LHUOIWVHXSPAQX-UHFFFAOYSA-N
2-4-fluorophenoxy phenyl methanol, 2-4-fluorophenoxy benzyl alcohol
3700253
[2-(4-fluorophenoxy)phenyl]methanol
C1=CC=C(C(=C1)CO)OC2=CC=C(C=C2)F

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
Quantity 250 mg
Formula Weight 218.23
Percent Purity 95%
Chemical Name or Material 2-(4-Fluorophenoxy)benzyl alcohol
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'