missing translation for 'onlineSavingsMsg'
Learn More

2,3-Dimethylthiophene, 97%, Thermo Scientific Chemicals

Catalog Number p-7030788
Click to view available options
Quantity:
1 g
5 g
This item is not returnable. View return policy
632-16-6
C6H8S
112.19
MFCD00130081
BZYUMXXOAYSFOW-UHFFFAOYSA-N
thiophene, 2,3-dimethyl, 2,3-dimethyl-thiophene, 2,3,dimethylthiophene, pubchem7751, 2.3-dimethylthiophene, dimethylthiophene, 2,3-dirnethylthiophene
34295
2,3-dimethylthiophene
CC1=C(C)C=CS1
This item is not returnable. View return policy
632-16-6
C6H8S
112.19
MFCD00130081
BZYUMXXOAYSFOW-UHFFFAOYSA-N
thiophene, 2,3-dimethyl, 2,3-dimethyl-thiophene, 2,3,dimethylthiophene, pubchem7751, 2.3-dimethylthiophene, dimethylthiophene, 2,3-dirnethylthiophene
34295
2,3-dimethylthiophene
CC1=C(C)C=CS1

It has also been found that 2,3-dimethylthiophene can be formylated to yield a dimethylthiophene aldehyde, which as expected proved to have the aldehyde group in the free a-position.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It has also been found that 2,3-dimethylthiophene can be formylated to yield a dimethylthiophene aldehyde, which as expected proved to have the aldehyde group in the free a-position.

Solubility
Insoluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents, water, moisture.
TRUSTED_SUSTAINABILITY
Boiling Point 142°C
Flash Point 26° (79°F)
Quantity 1 g
UN Number UN1993
Solubility Information Insoluble in water.
Formula Weight 112.19
Percent Purity 97%
Chemical Name or Material 2,3-Dimethylthiophene

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'