missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2,3-Difluorotoluene, 98%, Thermo Scientific™
Click to view available options
Quantity:
1 g
5 g
25 g
Specifications
Density | 1.0960g/mL |
Boiling Point | 119.0°C to 121.0°C |
Flash Point | 17°C |
Infrared Spectrum | Authentic |
Assay Percent Range | 97.5% min. (GC) |
Quantity | 5 g |
Specific Gravity | 1.096 |
Formula Weight | 128.12 |
Percent Purity | 98% |
Chemical Name or Material | 2,3-Difluorotoluene |
Safety and Handling
Product Identifier
- 2,3-Difluorotoluene, 98%
- Danger
- Acute toxicity Category 4
- Serious eye damage/eye irritation Category 1
- Flammable liquid Category 2
- Skin corrosion/irritation Category 2
- Specific target organ toxicity Category 3
- H318-Causes serious eye damage.
- H315-Causes skin irritation.
- H335-May cause respiratory irritation.
- H302-Harmful if swallowed.
- H225-Highly flammable liquid and vapour.
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P310-Immediately call a POISON CENTER/doctor/.
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'