Learn More
2,3-Difluoro-4-(trifluoromethyl)pyridine, 97%, Thermo Scientific™
Catalog Number p-4380085
Click to view available options
Quantity:
1 g
5 g
1215034-77-7
C6H2F5N
183.07
ZOMVJIOHXSXCIP-UHFFFAOYSA-N
2,3-difluoro-4-trifluoromethyl pyridine, alpha,alpha,alpha,2,3-pentafluoro-4-picoline
13491638
2,3-difluoro-4-(trifluoromethyl)pyridine
C1=CN=C(C(=C1C(F)(F)F)F)F
1215034-77-7
C6H2F5N
183.07
ZOMVJIOHXSXCIP-UHFFFAOYSA-N
2,3-difluoro-4-trifluoromethyl pyridine, alpha,alpha,alpha,2,3-pentafluoro-4-picoline
13491638
2,3-difluoro-4-(trifluoromethyl)pyridine
C1=CN=C(C(=C1C(F)(F)F)F)F
Boiling Point 82.0°C to 84.0°C (400.0 Torr)
Assay Percent Range 96% min. (GC)
Packaging Glass Bottle
Quantity 1 g
Formula Weight 183.07
Percent Purity 97%
Chemical Name or Material 2, 3-Difluoro-4-(trifluoromethyl)pyridine
compliance-icons compliance-icons
Product Identifier
  • 2,3-Difluoro-4-(trifluoromethyl)pyridine
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Flammable liquid Category 3
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H302-Harmful if swallowed.
  • H312-Harmful in contact with skin.
  • H332-Harmful if inhaled.
  • H226-Flammable liquid and vapour.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.