missing translation for 'onlineSavingsMsg'
Learn More

2,1,3-Benzothiadiazole-5-carboxylic acid, 97%, Thermo Scientific Chemicals

Catalog Number p-7031859
Click to view available options
Quantity:
250 mg
1 g
5 g
This item is not returnable. View return policy
16405-98-4
C7H4N2O2S
180.181
MFCD01647555
YHMXJZVGBCACMT-UHFFFAOYSA-N
benzo c 1,2,5 thiadiazole-5-carboxylic acid, benzo 1,2,5 thiadiazole-5-carboxylic acid, 2??,1,3-benzothiadiazole-5-carboxylic acid, benzo c 1,2,5-thiadiazole-5-carboxylic acid, 4ddm, 2,1,3-benzothiadiazole-5-carboxylicacid, benzo-2,1,3-thiadiazole-5-carboxylic acid, 2,1,3-benzothiadiazole-5-carboxylic acid, benzo 1,2, 5 thiadiazole-5-carboxylic acid, 2$l^ 4 ,1,3-benzothiadiazole-5-carboxylic acid
602011
2,1,3-benzothiadiazole-5-carboxylic acid
C1=CC2=NSN=C2C=C1C(=O)O
This item is not returnable. View return policy
16405-98-4
C7H4N2O2S
180.181
MFCD01647555
YHMXJZVGBCACMT-UHFFFAOYSA-N
benzo c 1,2,5 thiadiazole-5-carboxylic acid, benzo 1,2,5 thiadiazole-5-carboxylic acid, 2??,1,3-benzothiadiazole-5-carboxylic acid, benzo c 1,2,5-thiadiazole-5-carboxylic acid, 4ddm, 2,1,3-benzothiadiazole-5-carboxylicacid, benzo-2,1,3-thiadiazole-5-carboxylic acid, 2,1,3-benzothiadiazole-5-carboxylic acid, benzo 1,2, 5 thiadiazole-5-carboxylic acid, 2$l^ 4 ,1,3-benzothiadiazole-5-carboxylic acid
602011
2,1,3-benzothiadiazole-5-carboxylic acid
C1=CC2=NSN=C2C=C1C(=O)O

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Melting Point 228°C to 230°C
Quantity 250 mg
Formula Weight 180.19
Percent Purity 97%
Chemical Name or Material 2,1,3-Benzothiadiazole-5-carboxylic acid

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'