Learn More
(1S)-3-[3-(3-Isopropyl-5-methyl-4H-1,2,4-triazol-4-yl)-exo-8-azabicyclo[3.2.1]oct-8-yl]-1-phenyl-1-propanamine, TRC
Specifications
Specifications
| Chemical Name or Material | (1S)-3-[3-(3-Isopropyl-5-methyl-4H-1,2,4-triazol-4-yl)-exo-8-azabicyclo[3.2.1]oct-8-yl]-1-phenyl-1-propanamine |
| CAS | 376348-71-9 |
| Molecular Formula | C22 H33 N5 |
| Quantity | 5 mg |
| Synonym | (alphaS,3-exo)-3-[3-Methyl-5-(1-methylethyl)-4H-1,2,4-triazol-4-yl]-alpha-phenyl-8-azabicyclo[3.2.1]octane-8-propanamine, (alphaS,1alpha,3-exo,3beta,5alpha)-3-[3-Methyl-5-(1-methylethyl)-4H-1,2,4-triazol-4-yl]-alpha-phenyl-8-azabicyclo[3.2.1]octane-8-propanamine |
| InChI Formula | InChI=1S/C22H33N5/c1-15(2)22-25-24-16(3)27(22)20-13-18-9-10-19(14-20)26(18)12-11-21(23)17-7-5-4-6-8-17/h4-8,15,18-21H,9-14,23H2,1-3H3/t18-,19+,20-,21-/m0/s1 |
| SMILES | CC(C)c1nnc(C)n1[C@@H]2C[C@H]3CC[C@@H](C2)N3CC[C@H](N)c4ccccc4 |
| IUPAC Name | (1S)-3-[(1R,5S)-3-(3-methyl-5-propan-2-yl-1,2,4-triazol-4-yl)-8-azabicyclo[3.2.1]octan-8-yl]-1-phenylpropan-1-amine |
| Molecular Weight (g/mol) | 367.53 |
| Formula Weight | 367.2736 |