Learn More
Bochem™ 18/10 Stainless Steel Poly Spoons
Manufactured from 18/10 stainless steel
Brand:  Bochem™ 3414
Additional Details : Weight : 0.80000kg
 View more versions of this product
Product Code. 11895141
  • 123.00€ / Pack of 10
Estimated Shipment
04-03-2021
Add to basket
Description
Description
  • Spoon comes in three different types
  • Autoclavable
  • Made of 18/10 stainless steel
  • Multi-purpose spoon for a wide range of uses
Specifications
Specifications
1Each
Poly Spoon
35mm
8mm
18/10 Stainless Steel
Yes
Multi-purpose spoon for a wide range of uses
20mm
500 x 20mm
500mm