missing translation for 'onlineSavingsMsg'
Learn More

17alpha-Hydroxy-11-deoxycorticosterone-21-acetate, TRC

Product Code. 30137366
Change view
Click to view available options
Quantity:
50 mg
500 mg
1 g
Unit Size:
1g
500mg
50mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30137366 1 g 1g
30134372 500 mg 500mg
17993435 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30137366 Supplier Toronto Research Chemicals Supplier No. TRCH9348001G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 640-87-9
Molecular Formula C23H32O5
Molecular Weight (g/mol) 388.4972
Synonym Pregn-4-ene-3,20-dione, 21-(acetyloxy)-17-hydroxy-, Corticosterone, 11-deoxy-17-hydroxy-, 21-acetate (6CI), Pregn-4-ene-3,20-dione, 17,21-dihydroxy-, 21-acetate (8CI), 21-(Acetyloxy)-17-hydroxypregn-4-ene-3,20-dione, 11-Deoxycortisol 21-acetate, 17,21-Dihydroxypregn-4-ene-3,20-dione 21-acetate, 17-Hydroxy-21-acetoxyprogesterone, 17α-Hydroxy-11-deoxycorticosterone-21-acetate, 21-Acetoxy-17-hydroxypregn-4-ene-3,20-dione, 21-Acetoxy-17α-hydroxyprogesterone, 4-Pregnene-17α,21-diol-3,20-dione 21-acetate, Cortexolone 21-acetate, Cortodoxon 21-acetate, NSC 81196, Reichstein substance S 21-acetate, Reichstein-S-acetate, Reichstein's substance S 21-acetate, Reichstein's substance S acetate
IUPAC Name [2-[(8R,9S,10R,13S,14S,17R)-17-hydroxy-10,13-dimethyl-3-oxo-2,6,7,8,9,11,12,14,15,16-decahydro-1H-cyclopenta[a]phenanthren-17-yl]-2-oxoethyl] acetate
SMILES CC(=O)OCC(=O)[C@@]1(O)CC[C@H]2[C@@H]3CCC4=CC(=O)CC[C@]4(C)[C@H]3CC[C@]12C

Specifications

Quantity 1 g
InChI Formula InChI=1S/C23H32O5/c1-14(24)28-13-20(26)23(27)11-8-19-17-5-4-15-12-16(25)6-9-21(15,2)18(17)7-10-22(19,23)3/h12,17-19,27H,4-11,13H2,1-3H3/t17-,18+,19+,21+,22+,23+/m1/s1
Formula Weight 388.225
Chemical Name or Material 17a-Hydroxy-11-deoxycorticosterone-21-acetate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.