missing translation for 'onlineSavingsMsg'
Learn More

17-Hydroxy-3,20-dioxopregn-4-en-21-yl Acetate (Reichstein's Substance S 21-Acetate), Mikromol™

Product Code. 17913456
Change view
Click to view available options
Quantity:
25 mg
Unit Size:
25mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
17913456 25 mg 25mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 17913456 Supplier Mikromol Supplier No. MM0185.270025

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.

17-Hydroxy-3,20-dioxopregn-4-en-21-yl Acetate (Reichstein's Substance S 21-Acetate)
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 17a-Hydroxy-11-deoxycorticosterone-21-acetate
CAS 640-87-9
Analyte or Component Names 17a-Hydroxy-11-deoxycorticosterone-21-acetate
Shipping Condition Room Temperature
Molecular Formula C23H32O5
Certifications/Compliance ISO 9001
Synonym Pregn-4-ene-3,20-dione, 21-(acetyloxy)-17-hydroxy-, Corticosterone, 11-deoxy-17-hydroxy-, 21-acetate (6CI), Pregn-4-ene-3,20-dione, 17,21-dihydroxy-, 21-acetate (8CI), 21-(Acetyloxy)-17-hydroxypregn-4-ene-3,20-dione, 11-Deoxycortisol 21-acetate, 17,21-Dihydroxypregn-4-ene-3,20-dione 21-acetate, 17-Hydroxy-21-acetoxyprogesterone, 17α-Hydroxy-11-deoxycorticosterone-21-acetate, 21-Acetoxy-17-hydroxypregn-4-ene-3,20-dione, 21-Acetoxy-17α-hydroxyprogesterone, 4-Pregnene-17α,21-diol-3,20-dione 21-acetate, Cortexolone 21-acetate, Cortodoxon 21-acetate, NSC 81196, Reichstein substance S 21-acetate, Reichstein-S-acetate, Reichstein's substance S 21-acetate, Reichstein's substance S acetate, 17-Hydroxy-3,20-dioxopregn-4-en-21-yl Acetate, Hydrocortisone Impurity K
InChI Formula InChI=1S/C23H32O5/c1-14(24)28-13-20(26)23(27)11-8-19-17-5-4-15-12-16(25)6-9-21(15,2)18(17)7-10-22(19,23)3/h12,17-19,27H,4-11,13H2,1-3H3/t17-,18+,19+,21+,22+,23+/m1/s1
SMILES CC(=O)OCC(=O)[C@@]1(O)CC[C@H]2[C@@H]3CCC4=CC(=O)CC[C@]4(C)[C@H]3CC[C@]12C
IUPAC Name [2-[(8R,9S,10R,13S,14S,17R)-17-hydroxy-10,13-dimethyl-3-oxo-2,6,7,8,9,11,12,14,15,16-decahydro-1H-cyclopenta[a]phenanthren-17-yl]-2-oxoethyl] acetate
Molecular Weight (g/mol) 388.4972
Quantity 25 mg
Formula Weight 388.225 g/mol
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.