Learn More
KGW Isotherm™ 1600mL Dish-Shaped Dewar Flasks
Suitable for use as a hot or cold bath, cooling of small samples, tempering of round-bottom flasks or on a magnetic stirrer.
Brand: KGW Isotherm™ 1196
Additional Details : Weight : 0.80000kg
Description
- Easy handling
- Fully silvered
- Structured aluminum cover that is both antimagnetic and stainless
- Manufactured from borosilicate glass 3.3
Specifications
170 mm | |
Cooling of small samples | |
145mm | |
Borosilicate Glass (3.3) |
Dish-shaped | |
110mm | |
205mm | |
1600 mL |