Learn More
Helios Italquartz 1234 DIN Standard Covers with Ring Handle, Transparent
1234 DIN standard covers with ring handle constructed of transparent, durable, resistant, and chemically inert quartz glass.
Brand:  Helios Italquartz 64P00603
Additional Details : Weight : 0.20000kg
 View more versions of this product
Product Code. 15808094
Quantity Price
1 81.12€ / Each
10 75.55€ / Each
Estimated Shipment: 08-10-2021
Add to basket
Description
Description
  • Transparent
  • Include ring handle
  • Conform to 1234 DIN standard
  • Chemically inert
  • Excellent electrical insulation
  • Excellent strength and durability
  • Exceptional thermal stability compared to glass
Specifications
Specifications
Transparent
CR 43-52-63 Crucible
Quartz
1234 DIN Standard Covers with Ring Handle