Learn More
Hellma™ Suprasil™ Quartz 104B Semi-Micro Cell Cuvettes
Designed to hold samples for spectroscopic experiments and optical analysis. Hellma™ Suprasil™ Quartz 104B Semi-Micro Cell Cuvettes are supplied with a PTFE lid.
Brand: Hellma™ 104-B-10-40
Additional Details : Weight : 0.80000kg
Description
- Sold in pairs
- Very chemically resistant
- Maintains shape, even at high temperatures up to approximately 1000°C
- 80% transmission from 320 to 2500nm
Specifications
Semi-micro Cell | |
Quartz Suprasil | |
4mm | |
10 mm | |
45 mm |
1.4 mL | |
PTFE | |
1Each | |
200 to 2500 nm | |
12.5 mm |