missing translation for 'onlineSavingsMsg'
Learn More

1-Thieno[3,2-d]pyrimidin-4-ylpiperidine-4-carboxylic acid, 97%, Thermo Scientific™

Catalog Number p-3522413
Click to view available options
Quantity:
1 g
5 g
Packaging:
Amber Glass Bottle
This item is not returnable. View return policy
910037-25-1
C12H13N3O2S
263.32
MFCD09065023
VWNHOIKUCNKRDY-UHFFFAOYSA-N
1-thieno 3,2-d pyrimidin-4-ylpiperidine-4-carboxylic acid, 1-thieno 3,2-d pyrimidin-4-yl piperidine-4-carboxylic acid, 4-piperidinecarboxylicacid,1-thieno 3,2-d pyrimidin-4-yl, 1-thiopheno 2,3-e pyrimidin-4-ylpiperidine-4-carboxylic acid, 4-piperidinecarboxylicacid, 1-thieno 3,2-d pyrimidin-4-yl
24229705
1-thieno[3,2-d]pyrimidin-4-ylpiperidine-4-carboxylic acid
OC(=O)C1CCN(CC1)C1=C2SC=CC2=NC=N1
This item is not returnable. View return policy
910037-25-1
C12H13N3O2S
263.32
MFCD09065023
VWNHOIKUCNKRDY-UHFFFAOYSA-N
1-thieno 3,2-d pyrimidin-4-ylpiperidine-4-carboxylic acid, 1-thieno 3,2-d pyrimidin-4-yl piperidine-4-carboxylic acid, 4-piperidinecarboxylicacid,1-thieno 3,2-d pyrimidin-4-yl, 1-thiopheno 2,3-e pyrimidin-4-ylpiperidine-4-carboxylic acid, 4-piperidinecarboxylicacid, 1-thieno 3,2-d pyrimidin-4-yl
24229705
1-thieno[3,2-d]pyrimidin-4-ylpiperidine-4-carboxylic acid
OC(=O)C1CCN(CC1)C1=C2SC=CC2=NC=N1
CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 1 g
Formula Weight 263.32
Percent Purity 97%
Chemical Name or Material 1-Thieno[3, 2-d]pyrimidin-4-ylpiperidine-4-carboxylic acid
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'