Learn More
[1-(Pyrid-3-ylmethyl)piperid-4-yl]methanol, 97%, Thermo Scientific™
Catalog Number p-3586554
Click to view available options
Quantity:
250 mg
1 g
Packaging:
Amber Glass Bottle
934570-59-9
C12H18N2O
206.289
MFCD09879988
RAJQONMXWKPTTO-UHFFFAOYSA-N
1-pyrid-3-ylmethyl piperid-4-yl methanol, 1-pyridin-3-ylmethyl piperidin-4-yl methanol, 4-piperidinemethanol,1-3-pyridinylmethyl, 1-3-pyridylmethyl-4-piperidyl methan-1-ol, 4-piperidinemethanol, 1-3-pyridinylmethyl-, dihydrochloride
24229775
[1-(pyridin-3-ylmethyl)piperidin-4-yl]methanol
C1CN(CCC1CO)CC2=CN=CC=C2
934570-59-9
C12H18N2O
206.289
MFCD09879988
RAJQONMXWKPTTO-UHFFFAOYSA-N
1-pyrid-3-ylmethyl piperid-4-yl methanol, 1-pyridin-3-ylmethyl piperidin-4-yl methanol, 4-piperidinemethanol,1-3-pyridinylmethyl, 1-3-pyridylmethyl-4-piperidyl methan-1-ol, 4-piperidinemethanol, 1-3-pyridinylmethyl-, dihydrochloride
24229775
[1-(pyridin-3-ylmethyl)piperidin-4-yl]methanol
C1CN(CCC1CO)CC2=CN=CC=C2
CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 250 mg
Formula Weight 206.29
Percent Purity 97%
Chemical Name or Material [1-(Pyrid-3-ylmethyl)piperid-4-yl]methanol