missing translation for 'onlineSavingsMsg'
Learn More

1-Propanol, puriss. p.a., ACS Reagent, ≥99.5% (GC), Honeywell™ Riedel-de Haën™
BIOPHARMA

Catalog Number p-7120039
Click to view available options
Quantity:
4 x 2.5 L
6 x 1 L
Packaging:
Poly Bottle
71-23-8
C3H8O
60.10
MFCD00002941
BDERNNFJNOPAEC-UHFFFAOYSA-N
1-propanol, propanol, propyl alcohol, n-propanol, n-propyl alcohol, ethylcarbinol, 1-hydroxypropane, optal, osmosol extra, propylic alcohol
1031
CHEBI:28831
propan-1-ol
CCCO
71-23-8
C3H8O
60.10
MFCD00002941
BDERNNFJNOPAEC-UHFFFAOYSA-N
1-propanol, propanol, propyl alcohol, n-propanol, n-propyl alcohol, ethylcarbinol, 1-hydroxypropane, optal, osmosol extra, propylic alcohol
1031
CHEBI:28831
propan-1-ol
CCCO

Puriss. p.a., Reag. Ph. Eur.,≥99.5% (GC)

  • Honeywell Riedel-de Haen has over 200 years of history in producing high-purity chemicals
  • It is the leading brand offering High-Purity Solvents for dedicated applications.
TRUSTED_SUSTAINABILITY
Melting Point -127°C
Color Colorless
Density 0.804g/mL at 25°C
Boiling Point 97°C
Quantity 6 x 1 L
Explosion Limit 0.137
Flash Point 22°C (71.6°F)
Ignition Point 700°F
Odor Alcohol-like
Packaging Poly Bottle
Refractive Index n20/D 1.384
Linear Formula CH3CH2CH2OH
Vapor Pressure 14.9mmHg (20°C), 10mmHg (147°C)
UN Number UN1274
Beilstein 1098242
Solubility Information Soluble in water and most organic solvents
Formula Weight 60.1g/mol
Percent Purity ≥99.5% (GC)
Chemical Name or Material 1-Propanol
Show More Show Less
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'