missing translation for 'onlineSavingsMsg'
Learn More

1-(Phenylsulfonyl)-1H-indole-2-carboxylic acid, 97%, Thermo Scientific™

Catalog Number p-2694538
Click to view available options
Quantity:
1 g
5 g
This item is not returnable. View return policy
40899-93-2
C15H11NO4S
301.316
MFCD03086089
QIWDUGKJAHJRAE-UHFFFAOYSA-N
1-phenylsulfonyl-1h-indole-2-carboxylic acid, 1-benzenesulfonyl indole-2-carboxylic acid, 1-benzenesulfonyl-1h-indole-2-carboxylic acid, 1h-indole-2-carboxylicacid, 1-phenylsulfonyl
2776219
1-(benzenesulfonyl)indole-2-carboxylic acid
C1=CC=C(C=C1)S(=O)(=O)N2C3=CC=CC=C3C=C2C(=O)O
This item is not returnable. View return policy
40899-93-2
C15H11NO4S
301.316
MFCD03086089
QIWDUGKJAHJRAE-UHFFFAOYSA-N
1-phenylsulfonyl-1h-indole-2-carboxylic acid, 1-benzenesulfonyl indole-2-carboxylic acid, 1-benzenesulfonyl-1h-indole-2-carboxylic acid, 1h-indole-2-carboxylicacid, 1-phenylsulfonyl
2776219
1-(benzenesulfonyl)indole-2-carboxylic acid
C1=CC=C(C=C1)S(=O)(=O)N2C3=CC=CC=C3C=C2C(=O)O
CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 97%
Packaging Amber glass bottle
Quantity 1 g
Formula Weight 301.32
Percent Purity 97%
Chemical Name or Material 1-(Phenylsulfonyl)-1H-indole-2-carboxylic acid
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'