missing translation for 'onlineSavingsMsg'
Learn More

1-Methyl-6-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-1H-indole, 95%, Thermo Scientific™

Catalog Number p-3484819
Click to view available options
Quantity:
250 mg
1 g
Packaging:
Amber Glass Bottle
This item is not returnable. View return policy
884507-19-1
C15H20BNO2
257.14
MFCD08690255
AYJLGLUJQKZRDL-UHFFFAOYSA-N
1-methyl-6-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl-1h-indole, 1-methyl-6-tetramethyl-1,3,2-dioxaborolan-2-yl indole, 1-methyl-6-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl-indole, 1-methyl-6-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl indole, 1-methyl-1h-indole-6-boronic acid pinacol ester, 1-methyl-1h-indole-6-boronic acid,pinacol ester, 1-methyl-1h-indol-6-yl boronic acid pinacol ester, 4,4,5,5-tetramethyl-2-1-methylindol-6-yl-1,3,2-dioxaborolane, 1-methyl-6-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl-1-h-indole, 1-methyl-6-4,4,5,5-tetramethyl-1,3,2-dioxaborolane-2-yl-1h-indole
18525769
1-methyl-6-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)indole
CN1C=CC2=CC=C(C=C12)B1OC(C)(C)C(C)(C)O1
This item is not returnable. View return policy
884507-19-1
C15H20BNO2
257.14
MFCD08690255
AYJLGLUJQKZRDL-UHFFFAOYSA-N
1-methyl-6-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl-1h-indole, 1-methyl-6-tetramethyl-1,3,2-dioxaborolan-2-yl indole, 1-methyl-6-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl-indole, 1-methyl-6-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl indole, 1-methyl-1h-indole-6-boronic acid pinacol ester, 1-methyl-1h-indole-6-boronic acid,pinacol ester, 1-methyl-1h-indol-6-yl boronic acid pinacol ester, 4,4,5,5-tetramethyl-2-1-methylindol-6-yl-1,3,2-dioxaborolane, 1-methyl-6-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl-1-h-indole, 1-methyl-6-4,4,5,5-tetramethyl-1,3,2-dioxaborolane-2-yl-1h-indole
18525769
1-methyl-6-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)indole
CN1C=CC2=CC=C(C=C12)B1OC(C)(C)C(C)(C)O1
CAS Min % 94.5
CAS Max % 100.0
Assay Percent Range 95%
Packaging Amber Glass Bottle
Quantity 250 mg
Formula Weight 257.14
Percent Purity 95%
Chemical Name or Material 1-Methyl-6-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-1H-indole
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'