missing translation for 'onlineSavingsMsg'
Learn More

1-Chlorobutane, ReagentPlus™, 99%, Honeywell™

Catalog Number p-7120027
Click to view available options
Quantity:
1 L
2.5 L
Packaging:
Glass Bottle
This item is not returnable. View return policy
109-69-3
C4H9Cl
92.57
MFCD00001009
VFWCMGCRMGJXDK-UHFFFAOYSA-N
butyl chloride, n-butyl chloride, butane, 1-chloro, chlorure de butyle, nbc wormer, n-propylcarbinyl chloride, sure shot, butane, chloro, chlorobutane, n-chlorobutane
8005
1-chlorobutane
CCCCCl
This item is not returnable. View return policy
109-69-3
C4H9Cl
92.57
MFCD00001009
VFWCMGCRMGJXDK-UHFFFAOYSA-N
butyl chloride, n-butyl chloride, butane, 1-chloro, chlorure de butyle, nbc wormer, n-propylcarbinyl chloride, sure shot, butane, chloro, chlorobutane, n-chlorobutane
8005
1-chlorobutane
CCCCCl

Reagent Grade, 99%

  • ≥95% for general laboratory use.
TRUSTED_SUSTAINABILITY
Melting Point -123°C
Color Colorless
Density 0.886g/mL at 25°C
Boiling Point 78.8°C
Explosion Limit 0.101
Flash Point −12°C (10.4°F)
Ignition Point 860°F
Packaging Glass Bottle
Linear Formula CH3(CH2)3Cl
Refractive Index n20/D 1.402(lit.)
Quantity 1 L
UN Number UN1127
Beilstein 1730909
Vapor Pressure 80.1mmHg (78.4°C)
Formula Weight 92.57g/mol
Percent Purity 99%
Grade ReagentPlus™
Chemical Name or Material 1-Chlorobutane
Show More Show Less
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'