missing translation for 'onlineSavingsMsg'
Learn More

1-Bromo-3,5-dimethoxybenzene, 97%, Thermo Scientific Chemicals

Catalog Number p-4258797
Click to view available options
Quantity:
5 g
25 g
Packaging:
Glass bottle
This item is not returnable. View return policy
20469-65-2
C8H9BrO2
217.06
MFCD06200685
KRWRFIMBWRVMKE-UHFFFAOYSA-N
3,5-dimethoxybromobenzene, benzene, 1-bromo-3,5-dimethoxy, 1,3-dimethoxy-5-bromobenzene, 1-bromo-3,5-dimethoxy-benzene, 1-bromo-3,5-dimethox, 3,5-dimethoxy-bromobenzene, bromo-3,5-dimethoxybenzene, 3,5-dimethoxy bromobenzene, acmc-1cs59, ksc495e7l
639187
1-bromo-3,5-dimethoxybenzene
COC1=CC(OC)=CC(Br)=C1
This item is not returnable. View return policy
20469-65-2
C8H9BrO2
217.06
MFCD06200685
KRWRFIMBWRVMKE-UHFFFAOYSA-N
3,5-dimethoxybromobenzene, benzene, 1-bromo-3,5-dimethoxy, 1,3-dimethoxy-5-bromobenzene, 1-bromo-3,5-dimethoxy-benzene, 1-bromo-3,5-dimethox, 3,5-dimethoxy-bromobenzene, bromo-3,5-dimethoxybenzene, 3,5-dimethoxy bromobenzene, acmc-1cs59, ksc495e7l
639187
1-bromo-3,5-dimethoxybenzene
COC1=CC(OC)=CC(Br)=C1

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
CAS Min % 96.0
CAS Max % 100.0
Melting Point 63.6°C to 65.7°C
Color White to Yellow
Infrared Spectrum Authentic
Assay Percent Range 96% min. (GC)
Packaging Glass bottle
Quantity 5 g
Formula Weight 217.06
Percent Purity 97%
Physical Form Crystalline Powder or Crystals
Chemical Name or Material 1-Bromo-3, 5-dimethoxybenzene
Show More Show Less
compliance-icons
Product Identifier
  • 1-Bromo-3,5-dimethoxybenzene
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'