missing translation for 'onlineSavingsMsg'
Learn More

1-(4-chlorophenyl)ethanol, 98%, Thermo Scientific™

Catalog Number 11914661
Click to view available options
Quantity:
5 g
25 g
This item is not returnable. View return policy
3391-10-4
C8H9ClO
156.61
MVOSNPUNXINWAD-UHFFFAOYNA-N
boc-gly-osu, boc-glycine n-hydroxysuccinimide ester, boc-glycine-n-hydroxysuccinimide ester, 2,5-dioxopyrrolidin-1-yl 2-tert-butoxycarbonyl amino acetate, tert-butyl 2-2,5-dioxo-1-pyrrolidinyl oxy-2-oxoethyl carbamate, bocglyosu, 2,5-dioxopyrrolidin-1-yl n-tert-butoxycarbonyl glycinate
76926
(2,5-dioxopyrrolidin-1-yl) 2-[(2-methylpropan-2-yl)oxycarbonylamino]acetate
CC(O)C1=CC=C(Cl)C=C1
This item is not returnable. View return policy
3391-10-4
C8H9ClO
156.61
MVOSNPUNXINWAD-UHFFFAOYNA-N
boc-gly-osu, boc-glycine n-hydroxysuccinimide ester, boc-glycine-n-hydroxysuccinimide ester, 2,5-dioxopyrrolidin-1-yl 2-tert-butoxycarbonyl amino acetate, tert-butyl 2-2,5-dioxo-1-pyrrolidinyl oxy-2-oxoethyl carbamate, bocglyosu, 2,5-dioxopyrrolidin-1-yl n-tert-butoxycarbonyl glycinate
76926
(2,5-dioxopyrrolidin-1-yl) 2-[(2-methylpropan-2-yl)oxycarbonylamino]acetate
CC(O)C1=CC=C(Cl)C=C1
CAS Min % 97.5
CAS Max % 100.0
Color Colorless to Yellow
Density 1.1700g/mL
Boiling Point 136°C (2.7 kPa)
Flash Point >110°C
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (GC)
Quantity 5 g
Specific Gravity 1.17
Solubility Information Solubility in water: in water:(20°C) insoluble
Formula Weight 156.61
Percent Purity 98%
Physical Form Liquid
Chemical Name or Material 1-(4-chlorophenyl)ethanol
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'