Learn More
1,3-Dicyclohexylbenzimidazolium chloride, 95%, ACROS Organics™
Brand: Acros Organics 435090010
Additional Details : CAS : 1034449-15-4 Weight : 0.05100kg
Quantity | 1g |
---|
Chemical Identifiers
1034449-15-4 | |
318.89 | |
1,3-dicyclohexylbenzimidazolium chloride, 1,3-dicyclohexyl-1??,3-benzodiazol-1-ylium chloride, 1,3-dicyclohexyl-1h-benzimidazol-3-ium chloride, 1,3-dicyclohexyl-1h-benzo d imidazol-3-ium chloride, 1,3-dicyclohexyl-2h-1,3-benzodiazol-2-ylium chloride | |
1,3-dicyclohexylbenzimidazol-3-ium;chloride |
C19H27ClN2 | |
JACUDTKAZHCIPU-UHFFFAOYSA-M | |
16218267 | |
C1CCC(CC1)N2C=[N+](C3=CC=CC=C32)C4CCCCC4.[Cl-] |
Specifications
1, 3-Dicyclohexylbenzimidazolium chloride | |
Authentic | |
Tan to White | |
1g | |
94.0 | |
95% | |
1,3-dicyclohexylbenzimidazolium chloride, 1,3-dicyclohexyl-1??,3-benzodiazol-1-ylium chloride, 1,3-dicyclohexyl-1h-benzimidazol-3-ium chloride, 1,3-dicyclohexyl-1h-benzo d imidazol-3-ium chloride, 1,3-dicyclohexyl-2h-1,3-benzodiazol-2-ylium chloride | |
C1CCC(CC1)N2C=[N+](C3=CC=CC=C32)C4CCCCC4.[Cl-] | |
318.89 | |
318.89 | |
95% |
67.30 to 75.90% C, 8.25 to 9.31% N | |
Glass Bottle | |
209.9°C to 211.1°C | |
1034449-15-4 | |
100.0 | |
C19H27ClN2 | |
JACUDTKAZHCIPU-UHFFFAOYSA-M | |
1,3-dicyclohexylbenzimidazol-3-ium;chloride | |
16218267 | |
Powder |
Safety and Handling
- 1,3-Dicyclohexylbenzimidazolium chloride
Signal Word
- Warning
Hazard Category
- Serious eye damage/eye irritation Category 2
- Skin corrosion/irritation Category 2
- Specific target organ toxicity Category 3
Hazard Statement
- H319-Causes serious eye irritation.
- H335-May cause respiratory irritation.
- H315-Causes skin irritation.
Precautionary Statement
- P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
- P302+P352-IF ON SKIN: Wash with plenty of water/.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.