missing translation for 'onlineSavingsMsg'
Learn More

1,3-Dichloro-2-propanol, 99%, Thermo Scientific Chemicals

Catalog Number p-105857
Click to view available options
Quantity:
250 mL
1 L
Packaging:
Glass bottle
This item is not returnable. View return policy
Regulatory: This product is an isolated intermediate transported in accordance with Regulation (EC) 1907/2006 (REACH). All customers must complete a declaration before delivery.
96-23-1
C3H6Cl2O
128.98
MFCD00000951
DEWLEGDTCGBNGU-UHFFFAOYSA-N
1,3-dichloro-2-propanol, dichlorohydrin, 1,3-dichloroisopropanol, 1,3-dichlorohydrin, glycerol 1,3-dichlorohydrin, 2-propanol, 1,3-dichloro, enodrin, 1,3-dichloro-2-hydroxypropane, 1,3-dichloroisopropyl alcohol, sym-glycerol dichlorohydrin
7289
CHEBI:18917
1,3-dichloropropan-2-ol
OC(CCl)CCl
This item is not returnable. View return policy
Regulatory: This product is an isolated intermediate transported in accordance with Regulation (EC) 1907/2006 (REACH). All customers must complete a declaration before delivery.
96-23-1
C3H6Cl2O
128.98
MFCD00000951
DEWLEGDTCGBNGU-UHFFFAOYSA-N
1,3-dichloro-2-propanol, dichlorohydrin, 1,3-dichloroisopropanol, 1,3-dichlorohydrin, glycerol 1,3-dichlorohydrin, 2-propanol, 1,3-dichloro, enodrin, 1,3-dichloro-2-hydroxypropane, 1,3-dichloroisopropyl alcohol, sym-glycerol dichlorohydrin
7289
CHEBI:18917
1,3-dichloropropan-2-ol
OC(CCl)CCl

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
CAS Min % 98.5
CAS Max % 100.0
Melting Point -4.0°C
Color Colorless to Yellow
Density 1.3510g/mL
Boiling Point 172.0°C to 176.0°C
Flash Point 74°C
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (GC)
Packaging Glass bottle
Linear Formula ClCH2CH(OH)CH2Cl
Refractive Index 1.4820 to 1.4840
Quantity 250 mL
Beilstein 01, 364
Merck Index 15, 3084
Specific Gravity 1.351
Solubility Information Solubility in water: 13g/L (20°C). Other solubilities: miscible with most common organic solvents
Viscosity 13.9 mPa.s (20°C)
Formula Weight 128.99
Percent Purity 99%
Physical Form Liquid
Chemical Name or Material 1, 3-Dichloro-2-propanol, 99%
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • 1,3-Dichloro-2-propanol
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 3
  • Acute toxicity Category 4
  • Carcinogenicity Category 1B
Hazard Statement
  • H350-May cause cancer.
  • H312-Harmful in contact with skin.
  • H301-Toxic if swallowed.
Precautionary Statement
  • P201-Obtain special instructions before use.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
  • P302+P352-IF ON SKIN: Wash with plenty of water/.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'