missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Alfa Aesar™ 1,2-Dihydronaphthalene, 96%
Click to view available options
Quantity:
5g
Unit Size:
5g
Description
Applications
1,2-Dihydronaphthalene is used as a pharmaceutical intermediate.
Solubility
Not miscible or difficult to mix with water.
Notes
Keep container tightly sealed. Store in cool, dry conditions in well sealed containers. Incompatible with oxidizing agents.
1,2-Dihydronaphthalene is used as a pharmaceutical intermediate.
Solubility
Not miscible or difficult to mix with water.
Notes
Keep container tightly sealed. Store in cool, dry conditions in well sealed containers. Incompatible with oxidizing agents.
Specifications
Specifications
| CAS | 447-53-0 |
| Boiling Point | 88°C to 89°C (16mmHg) |
| Flash Point | 74°C (165°F) |
| Molecular Formula | C10H10 |
| MDL Number | MFCD00001672 |
| Quantity | 5g |
| Beilstein | 1851372 |
| Synonym | dialin, 1,2-dialin, diolin, delta1-dialin, dihydronaphthalene, naphthalene, 1,2-dihydro, dialin van, delta 1-dialin, dihydronapthalene, naphthalene, dihydro |
| InChI Key | KEIFWROAQVVDBN-UHFFFAOYSA-N |
| SMILES | C1CC2=CC=CC=C2C=C1 |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction