missing translation for 'onlineSavingsMsg'
Learn More

1,1'-Bis((2R,5R)-2,5-diisopropylphospholano)ferrocene(cyclooctadiene)rhodium(I) tetrafluoroborate, 97%, Thermo Scientific™

Catalog Number p-4258766
Click to view available options
Quantity:
100 mg
250 mg
Packaging:
Glass bottle
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
849773-97-3
C38H60BF4FeP2Rh
824.40
MFCD15144872
MFSWTFGIDMKBOY-UHFFFAOYNA-N
1,1 inverted exclamation marka-bis 2r,5r-2,5-diisopropylphospholano ferrocene cyclooctadiene rhodium i tetrafluoroborate
71310759
(1Z,5Z)-cycloocta-1,5-diene;(2R,5R)-1-cyclopenta-2,4-dien-1-yl-2,5-di(propan-2-yl)phospholane;iron(2+);rhodium;tetrafluoroborate
[Fe].[Rh+].F[B-](F)(F)F.C1C\C=C/CC\C=C/1.CC(C)[C@H]1CC[C@H](C(C)C)P1c1cccc1.CC(C)[C@H]1CC[C@H](C(C)C)P1c1cccc1
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
849773-97-3
C38H60BF4FeP2Rh
824.40
MFCD15144872
MFSWTFGIDMKBOY-UHFFFAOYNA-N
1,1 inverted exclamation marka-bis 2r,5r-2,5-diisopropylphospholano ferrocene cyclooctadiene rhodium i tetrafluoroborate
71310759
(1Z,5Z)-cycloocta-1,5-diene;(2R,5R)-1-cyclopenta-2,4-dien-1-yl-2,5-di(propan-2-yl)phospholane;iron(2+);rhodium;tetrafluoroborate
[Fe].[Rh+].F[B-](F)(F)F.C1C\C=C/CC\C=C/1.CC(C)[C@H]1CC[C@H](C(C)C)P1c1cccc1.CC(C)[C@H]1CC[C@H](C(C)C)P1c1cccc1
CAS Min % 96.0
CAS Max % 100.0
Melting Point 235.0°C to 238.0°C
Color Yellow to Brown
Infrared Spectrum Authentic
Assay Percent Range 97%
Packaging Glass bottle
Quantity 100 mg
Solubility Information Solubility in water: insoluble.
Formula Weight 824.39
Percent Purity 97%
Physical Form Crystalline Powder or Crystals
Chemical Name or Material 1,1′-Bis((2R,5R)-2,5-diisopropylphospholano)ferrocene(cyclooctadiene)rhodium(I) tetrafluorobor
Show More Show Less
compliance-icons
Product Identifier
  • 1,1'-Bis((2R,5R)-2,5-diisopropylphospholano)ferrocene(cyclooctadiene)rhodium(I) tetrafluoroborate
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'