accessibility menu, dialog, popup

UserName

CAS RN 81335-37-7

CH3H3CH3CNNHONHOO

CAS RN 81335-37-7

IUPAC Name: 2-[4-methyl-5-oxo-4-(propan-2-yl)-4,5-dihydro-1H-imidazol-2-yl]quinoline-3-carboxylic acid
Synonyms: imazaquin
Molecular Weight (g/mol): 311.34
Molecular Formula: C17H17N3O3
InChi Key: CABMTIJINOIHOD-UHFFFAOYNA-N
SMILES: CC(C)C1(C)N=C(NC1=O)C1=C(C=C2C=CC=CC2=N1)C(O)=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results