accessibility menu, dialog, popup

UserName

CAS RN 7783-59-7

FHPbFHFHHF

CAS RN 7783-59-7

IUPAC Name:
Synonyms:
Molecular Weight (g/mol): 287.2
Molecular Formula: F4H4Pb
InChi Key: KKUOLSVLJAIIPH-UHFFFAOYSA-N
SMILES: F.F.F.F.[Pb]

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results