CAS RN 7782-40-3
CAS RN 7782-40-3
IUPAC Name:
carbon
Synonyms:
carbon kohlenstoff atomic carbon carbonium carbone carbono carbon atom
Molecular Weight (g/mol):
12.01
Molecular Formula:
C
InChi Key:
OKTJSMMVPCPJKN-UHFFFAOYSA-N
SMILES:
[C]
1
–
1
of
1
results
1
Diamond powder, synthetic, 40-60 micron, 99.9% (metals basis)
Diamond powder is generally used as an abrasive to grind and polish hard surfaces | CAS: 7782-40-3