accessibility menu, dialog, popup

UserName

CAS RN 77156-85-5

CH3OONClOCH3

CAS RN 77156-85-5

IUPAC Name: ethyl 4-chloro-7-methoxyquinoline-3-carboxylate
Synonyms: ethyl 4-chloro-7-methoxyquinoline-3-carboxylate
Molecular Weight (g/mol): 265.69
Molecular Formula: C13H12ClNO3
InChi Key: IXMRUGGFGJIANG-UHFFFAOYSA-N
SMILES: CCOC(=O)C1=CN=C2C=C(OC)C=CC2=C1Cl

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results