accessibility menu, dialog, popup

UserName

CAS RN 77-40-7

H3CCH3OHOH

CAS RN 77-40-7

IUPAC Name: 4-[2-(4-hydroxyphenyl)butan-2-yl]phenol
Synonyms: bisphenol B
Molecular Weight (g/mol): 242.32
Molecular Formula: C16H18O2
InChi Key: HTVITOHKHWFJKO-UHFFFAOYSA-N
SMILES: CCC(C)(C1=CC=C(O)C=C1)C1=CC=C(O)C=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results