accessibility menu, dialog, popup

UserName

CAS RN 76903-88-3

FFClO

CAS RN 76903-88-3

IUPAC Name: 3,4-difluorobenzoyl chloride
Synonyms: 3,4-difluorobenzoyl chloride
Molecular Weight (g/mol): 176.55
Molecular Formula: C7H3ClF2O
InChi Key: RPQWXGVZELKOEU-UHFFFAOYSA-N
SMILES: FC1=CC=C(C=C1F)C(Cl)=O
Boiling Point 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results