CAS RN 7689-03-4
CAS RN 7689-03-4
IUPAC Name:
(19S)-19-ethyl-19-hydroxy-17-oxa-3,13-diazapentacyclo[11.8.0.0²,¹¹.0⁴,⁹.0¹⁵,²⁰]henicosa-1(21),2(11),3,5,7,9,15(20)-heptaene-14,18-dione
Synonyms:
camptothecin camptothecin (+ )- (.+-.)-camptothecin (+)-camptothecin (+)-camptothecine 20(S)-camptothecin camptothecine D-camptothecin 20(s)-camptothecine CPT
Molecular Weight (g/mol):
348.36
Molecular Formula:
C20H16N2O4
InChi Key:
VSJKWCGYPAHWDS-FQEVSTJZSA-N
SMILES:
CC[C@@]1(O)C(=O)OCC2=C1C=C1N(CC3=C1N=C1C=CC=CC1=C3)C2=O
1
–
3
of
3
results
1
Thermo Scientific Chemicals Camptothecin
CAS: 7689-03-4 Synonym: (S)-(+)-Camptothecin
2
Promotions Available
(+)-Camptothecin, 98%
(+)-Camptothecin, 98%
CAS: 7689-03-4 Molecular Formula: C20H16N2O4 Molecular Weight (g/mol): 348.35 MDL Number: MFCD00081076 InChI Key: VSJKWCGYPAHWDS-FQEVSTJZSA-N Synonym: camptothecin,camptothecine,s-+-camptothecin,campathecin,+-camptothecine,d-camptothecin,20 s-camptothecine,+-camptothecin,21,22-secocamptothecin-21-oic acid lactone,s-camptothecin PubChem CID: 24360 ChEBI: CHEBI:27656 SMILES: CCC1(C2=C(COC1=O)C(=O)N3CC4=CC5=CC=CC=C5N=C4C3=C2)O