accessibility menu, dialog, popup

UserName

CAS RN 420831-40-9

NSOOONHBr

CAS RN 420831-40-9

IUPAC Name: 3-(azepane-1-sulfonyl)-N-(3-bromophenyl)benzamide
Synonyms: 3-(azepane-1-sulfonyl)-N-(3-bromophenyl)benzamide
Molecular Weight (g/mol): 437.35
Molecular Formula: C19H21BrN2O3S
InChi Key: IYAYHZZWYNXHEQ-UHFFFAOYSA-N
SMILES: BrC1=CC=CC(NC(=O)C2=CC(=CC=C2)S(=O)(=O)N2CCCCCC2)=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results