accessibility menu, dialog, popup

UserName

CAS RN 319-88-0

FClFClFCl

CAS RN 319-88-0

IUPAC Name: 1,3,5-trichloro-2,4,6-trifluorobenzene
Synonyms: 1,3,5-trichloro-2,4,6-trifluorobenzene
Molecular Weight (g/mol): 235.41
Molecular Formula: C6Cl3F3
InChi Key: QPXZZPSKCVNHFW-UHFFFAOYSA-N
SMILES: FC1=C(Cl)C(F)=C(Cl)C(F)=C1Cl
Boiling Point 
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results