accessibility menu, dialog, popup

UserName

CAS RN 1449132-61-9

BFOHNNOOClHOOH

CAS RN 1449132-61-9

IUPAC Name: {3-[(2-chloro-4-nitrophenyl)carbamoyl]-5-fluorophenyl}boronic acid
Synonyms: 3-[(2-chloro-4-nitrophenyl)carbamoyl]-5-fluorophenylboronic acid
Molecular Weight (g/mol): 338.48
Molecular Formula: C13H9BClFN2O5
InChi Key: JGVQEEYFVVEKHQ-UHFFFAOYSA-N
SMILES: OB(O)C1=CC(=CC(F)=C1)C(=O)NC1=C(Cl)C=C(C=C1)[N+]([O-])=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results