accessibility menu, dialog, popup

UserName

CAS RN 126766-32-3

H3COONNNOClClOHOOHO(E)

CAS RN 126766-32-3

IUPAC Name: (2E)-but-2-enedioic acid; methyl 4-[2-(3,4-dichlorophenyl)acetyl]-3-[(pyrrolidin-1-yl)methyl]piperazine-1-carboxylate
Synonyms: fumaric acid; methyl 4-[2-(3,4-dichlorophenyl)acetyl]-3-(pyrrolidin-1-ylmethyl)piperazine-1-carboxylate
Molecular Weight (g/mol): 530.4
Molecular Formula: C23H29Cl2N3O7
InChi Key: ABTNETSDXZBJTE-WLHGVMLRNA-N
SMILES: OC(=O)\C=C\C(O)=O.COC(=O)N1CCN(C(CN2CCCC2)C1)C(=O)CC1=CC=C(Cl)C(Cl)=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results