CAS RN 107549-26-8
CAS RN 107549-26-8
IUPAC Name:
magnesium(2+) (2,6-difluorophenyl)methanide bromide
Synonyms:
magnesium(2+) (2,6-difluorophenyl)methanide bromide
Molecular Weight (g/mol):
231.32
Molecular Formula:
C7H5BrF2Mg
InChi Key:
KDVWBIWSDJCATG-UHFFFAOYSA-M
SMILES:
[Mg++].[Br-].[CH2-]C1=C(F)C=CC=C1F
1
–
1
of
1
results